Abnova
Manufacturer Code:MAB7972
Catalog # 89-113-256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F39P7F11 |
Immunogen | A synthetic peptide (conjugated with ovalbumin) corresponding to human CCT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of CCT2.
For Research Use Only
Protein Aliases: 99D8.1; CCT-beta; CCTB; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1, subunit 2 (beta); epididymis secretory sperm binding protein Li 100n; MGC142074; MGC142076; PRO1633; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; TCP-1-beta
Gene Aliases: 99D8.1; CCT-beta; CCT2; CCTB; HEL-S-100n; PRO1633; TCP-1-beta
UniProt ID: (Human) P78371
Entrez Gene ID: (Human) 10576
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.