Abnova
Manufacturer Code:H00010576K
Catalog # 89-201-689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CCT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CCT2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 99D8.1 CCT-beta CCTB MGC142074 MGC142076 PRO1633 TCP-1-beta; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1, subunit 2 (beta); epididymis secretory sperm binding protein Li 100n; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; TCP-1-beta
Gene Aliases: 99D8.1; CCT-beta; CCT2; CCTB; HEL-S-100n; PRO1633; TCP-1-beta
UniProt ID: (Human) P78371
Entrez Gene ID: (Human) 10576
Molecular Function:
chaperone
chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.