Invitrogen
Manufacturer Code:PA578951
Catalog # PIPA578951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human CCS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Copper chaperone for superoxide dismutase; Copper chaperone for superoxide dismutase Superoxide dismutase copper chaperone CCS; Superoxide dismutase copper chaperone
Gene Aliases: CCS; Ccsd
UniProt ID: (Human) O14618, (Rat) Q9JK72, (Mouse) Q9WU84
Entrez Gene ID: (Human) 9973, (Rat) 84485, (Mouse) 12460
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.