Invitrogen
Manufacturer Code:MA524209
Catalog # PIMA524209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 498321 |
Immunogen | RBL-1 rat basophilic leukemia cell line transfected with mouse CCRL2/LCCR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C chemokine receptor-like 2; CCRL2 CCR11 Chemokine receptor X CRAM CRAM-A CRAM-B CKRX C-C chemokine receptor-like 2 Chemokine receptor CCR11 HCR; Chemokine receptor CCR11; G-protein coupled beta chemokine receptor; L-CCR; Lipopolysaccharide-inducible C-C chemokine receptor
Gene Aliases: 1810047I05Rik; Ackr5; CCR11; Ccrl2; Cmkbr1l2; E01; L-CCR; Lccr
UniProt ID: (Mouse) O35457
Entrez Gene ID: (Mouse) 54199
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.