Invitrogen
Manufacturer Code:PA540819
Catalog # PIPA540819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CCRL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: atypical chemokine receptor 5; C-C chemokine receptor-like 2; C-C motif chemokine receptor like 2; chemokine (C-C motif) receptor-like 2; Chemokine (C-C motif) receptor-like 2 CKRX CRAM-A CRAM-B HCR MGC116710 CRAM; Chemokine C-C motif receptor-like 2; chemokine C-C motif receptor-like 2-like; Chemokine receptor CCR11; Chemokine receptor X; G-protein coupled beta chemokine receptor; L-CCR; Lipopolysaccharide-inducible C-C chemokine receptor; Putative MCP-1 chemokine receptor
Gene Aliases: 1810047I05Rik; ACKR5; CCR11; CCR6; CCRL2; CKRX; Cmkbr1l2; CRAM; CRAM-A; CRAM-B; E01; HCR; L-CCR; Lccr
UniProt ID: (Human) O00421, (Bovine) Q0II78, (Mouse) O35457
Entrez Gene ID: (Human) 9034, (Bovine) 515976, (Horse) 100054419, (Rabbit) 100348482, (Rat) 316019, (Mouse) 54199, (Guinea pig) 100719164
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.