Invitrogen
Manufacturer Code:PA533411
Catalog # PIPA533411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human CCRL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: atypical chemokine receptor 5; C-C chemokine receptor-like 2; CCRL2 CCR11 Chemokine receptor X CRAM CRAM-A CRAM-B CKRX C-C chemokine receptor-like 2 Chemokine receptor CCR11 HCR; chemokine (C-C motif) receptor-like 2; Chemokine receptor CCR11; Chemokine receptor X; Putative MCP-1 chemokine receptor
Gene Aliases: ACKR5; CCR11; CCR6; CCRL2; CKRX; CRAM; CRAM-A; CRAM-B; HCR
UniProt ID: (Human) O00421
Entrez Gene ID: (Human) 9034
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.