Invitrogen
Manufacturer Code:PA528743
Catalog # PIPA528743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human CCRL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: atypical chemokine receptor 5; C-C chemokine receptor-like 2; chemokine (C-C motif) receptor-like 2; Chemokine receptor CCR11; Chemokine receptor X; CKRX CRAM CRAM-A CRAM-B HCR C-C chemokine receptor-like 2 chemokine receptor CCR11 chemokine receptor X putative MCP-1 chemokine receptor; G-protein coupled beta chemokine receptor; L-CCR; Lipopolysaccharide-inducible C-C chemokine receptor; Putative MCP-1 chemokine receptor
Gene Aliases: 1810047I05Rik; ACKR5; CCR11; CCR6; CCRL2; CKRX; Cmkbr1l2; CRAM; CRAM-A; CRAM-B; E01; HCR; L-CCR; Lccr
UniProt ID: (Human) O00421, (Mouse) O35457
Entrez Gene ID: (Human) 9034, (Mouse) 54199
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.