Invitrogen
Manufacturer Code:PA121612
Catalog # PIPA121612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide: SSSIYDYDYLDDMTILVCRKDEVLSFGRC corresponding to amino acids 18-45 of Mouse beta Chemokine Receptor D6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Atypical chemokine receptor 2; beta Chemokine Receptor D6; C-C chemokine receptor D6; chemokine binding protein 2; chemokine decoy receptor D6; Chemokine-binding protein 2; Chemokine-binding protein D6; D6 beta-chemokine receptor
Gene Aliases: Ackr2; AI464239; Ccbp2; CCR10; CCR9; Cmkbr9; D6
UniProt ID: (Mouse) O08707
Entrez Gene ID: (Mouse) 59289
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.