R&D Systems
Manufacturer Code:FAB197A100
Catalog # FAB197A100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 150503 |
Immunogen | Human CCR7 transfectants Met1-Pro378 Accession # AAA58615 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: BLR2; BLR2 BLR2C-C chemokine receptor type 7 C-C CKR-7 CC-CKR-7 CCR7 CCR-7 CD197 CD197 antigen CDw197 CDw197CC chemokine receptor 7 chemokine (C-C motif) receptor 7 CMKBR7 CMKBR7chemokine (C-C) receptor 7 EBI1 EBI1EBV-induced G protein-coupled receptor 1 EBV-induced G-protein coupled receptor 1 Epstein-Barr virus induced gene 1 Epstein-Barr virus induced G-protein coupled receptor Epstein-Barr virus-induced G-protein coupled receptor 1 EVI1 lymphocyte-specific G protein-coupled peptide receptor MIP-3 beta receptor; Bukitt's lymphoma receptor 2; C-C chemokine receptor type 7; C-C CKR-7; CC chemokine receptor 7; CD197; CDw197; chemokine (C-C motif) receptor 7; EBI1; EBV-induced G protein-coupled receptor 1; Epstein-Barr virus induced gene 1; Epstein-Barr virus-induced G-protein coupled receptor 1; lymphocyte-specific G protein-coupled peptide receptor; MIP-3 beta receptor
Gene Aliases: BLR2; CC-CKR-7; CCR-7; CCR7; CD197; CDw197; CMKBR7; EBI1; EVI1
UniProt ID: (Human) P32248
Entrez Gene ID: (Human) 1236
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.