R&D Systems
Manufacturer Code:FAB590G025
Catalog # FAB590G025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 140706 |
Immunogen | RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6 Met1-Met367 Accession # O54689 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat Monoclonal Antibody
Protein Aliases: BN-1 CC-CKR-6 CCR6 CCR-6 CD196 CD196 antigen chemokine (C-C motif) receptor 6 CKRL3 CKR-L3 CKR-L3G-protein coupled receptor 29 CKRL3LARC receptor CMKBR6 CMKBR6STRL22 DRY6 GPR29 GPR29C-C CKR-6 GPRCY4 GPR-CY4 GPRCY4DRY-6 lymphocyte 22 STRL22; C-C chemokine receptor type 6; C-C CKR-6; CD196; chemokine (C-C motif) receptor 6; chemokine (C-C) receptor 6; Chemokine receptor-like 3; CKR-L3; DRY6; G protein-coupled receptor 29; G-protein coupled receptor 29; GPR-CY4; GPRCY4; LARC receptor; seven-transmembrane receptor, lymphocyte, 22
Gene Aliases: BN-1; C-C CKR-6; CC-CKR-6; CCR-6; CCR6; CD196; CKR-L3; CKRL3; CMKBR6; DCR2; DRY6; GPR29; GPRCY4; STRL22
UniProt ID: (Human) P51684
Entrez Gene ID: (Human) 1235
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.