R&D Systems
Manufacturer Code:FAB1802P100
Catalog # FAB1802P100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CTC5 |
Immunogen | CHO Chinese hamster ovary cell line transfected with human CCR5 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: C-C chemokine receptor type 5; C-C CKR-5; C-C CKR-5 C-C motif chemokine receptor 5 A159A CCCKR5 CC-CKR-5FLJ78003 CCR5 CCR-5 CD195 CD195 antigen chemokine (C-C motif) receptor 5 chemokine receptor CCR5 chemr13 CKR5 CKR-5 HIV-1 fusion coreceptor IDDM22CMKBR5C-C chemokine receptor type 5; C-C motif chemokine receptor 5 A159A; CD195; chemokine (C-C motif) receptor 5; chemokine receptor CCR5; CHEMR13; HIV-1 fusion coreceptor
Gene Aliases: CC-CKR-5; CCCKR5; CCR-5; CCR5; CD195; CKR-5; CKR5; CMKBR5; IDDM22
UniProt ID: (Human) P51681
Entrez Gene ID: (Human) 1234
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.