Proteintech
Manufacturer Code:161541AP150UL
Catalog # 50-173-0758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C C chemokine receptor type 2 C C CKR 2 CC CKR 2 CCR 2 CCR2 CCR2a CCR2b CD192 CKR2 CKR2A CKR2B CMKBR2 MCP 1 R; C-C chemokine receptor type 2; C-C CKR-2; CD192; chemokine (C-C motif) receptor 2; MCP-1 receptor; MCP-1-R; Monocyte chemoattractant protein 1 receptor; monocyte chemotactic protein 1 receptor
Gene Aliases: CC-CKR-2; CCR-2; CCR2; CCR2A; CCR2B; CD192; CKR2; CKR2A; CKR2B; CMKBR2; MCP-1-R
UniProt ID: (Human) P41597
Entrez Gene ID: (Human) 729230
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.