Invitrogen
Manufacturer Code:PA523045
Catalog # PIPA523045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an N-terminal portion of the mouse CCR2 protein (within residues 20-100). |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C chemokine receptor type 2; C-C CHEMOKINE RECEPTOR TYPE 2 (C-C CKR-2) (CC-CKR-2) (CCR-2) (CCR2) (JE/FIC RECEPTOR) (MCP-1 RECEPTOR); C-C CKR-2; CCR-2; CD192; chemoattractant protein-1 receptor; chemokine (C-C motif) receptor 2; chemokine (C-C) receptor 2; hCG&lowbar14621 CC-CKR-2 CCR2A CCR2B CD192 CKR2 CKR2A CKR2B CMKBR2 MCP-1-R C-C CKR-2 C-C chemokine receptor type 2 CCR-2 MCP-1 receptor monocyte chemoattractant protein 1 receptor monocyte chemotactic protein 1 receptor; JE/FIC receptor; MCP-1 receptor; MCP-1-R; MIP-1 alphaR; Monocyte chemoattractant protein 1 receptor; monocyte chemotactic protein 1 receptor
Gene Aliases: CC-CKR-2; CCR-2; CCR2; CCR2A; CCR2B; CD192; CKR2; CKR2A; CKR2B; CMKBR2; MCP-1-R; mJe-r
UniProt ID: (Human) P41597, (Mouse) P51683
Entrez Gene ID: (Human) 729230, (Mouse) 12772
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.