R&D Systems
Manufacturer Code:FAB145P025
Catalog # FAB145P025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 53504 |
Immunogen | Swiss 3T3 mouse cell line transfected with human CCR1 Accession # P32246 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: C-C chemokine receptor type 1; C-C chemokine receptor type 1 CC-CKR-1 CCR1 CCR-1 CD191 CD191 antigen chemokine (C-C motif) receptor 1 CKR1 CKR-1 CMKBR1SCYAR1 CMKR1 HM145C-C CKR-1 LD78 receptor Macrophage inflammatory protein 1-alpha receptor MIP-1alpha-R MIP1aR RANTES receptor RANTES-R; C-C CKR-1; CC-CKR-1; CCR-1; CD191; chemokine (C-C motif) receptor 1; HM145; LD78 receptor; Macrophage inflammatory protein 1-alpha receptor; MIP-1alpha-R; RANTES receptor; RANTES-R
Gene Aliases: CCR1; CD191; CKR-1; CKR1; CMKBR1; CMKR1; HM145; MIP1aR; SCYAR1
UniProt ID: (Human) P32246
Entrez Gene ID: (Human) 1230
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.