Abnova
Manufacturer Code:PAB26120
Catalog # 89-127-762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acids at N-terminus of human CCR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CCR1.
For Research Use Only
Protein Aliases: C-C chemokine receptor type 1; C-C CKR-1; CC-CKR-1; CCR-1; CD191; chemokine (C-C motif) receptor 1; CKR-1; CKR1; CMKBR1; HM145; LD78 receptor; Macrophage inflammatory protein 1-alpha receptor; MIP-1alpha-R; MIP1aR; RANTES receptor; RANTES-R; SCYAR1
Gene Aliases: CCR1; CD191; CKR-1; CKR1; CMKBR1; CMKR1; HM145; MIP1aR; SCYAR1
UniProt ID: (Human) P32246
Entrez Gene ID: (Human) 1230
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.