Invitrogen
Manufacturer Code:PA534380
Catalog # PIPA534380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid synthetic peptide near the carboxy terminus of human CCP110 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCP110 Centriolar coiled coil protein 110 Centriolin CNTRL FAN CEP1 CEP110; centriolar coiled coil protein 110kDa; centriolar coiled-coil protein 110kDa; Centriolar coiled-coil protein of 110 kDa; centrosomal protein CP110; Centrosomal protein of 110 kDa; Cep110; CP110
Gene Aliases: 6330503K22Rik; AA415922; AI427129; AW557948; CCP110; CEP110; CP110; KIAA0419; RGD1310509
UniProt ID: (Human) O43303, (Mouse) Q7TSH4
Entrez Gene ID: (Human) 9738, (Mouse) 101565, (Rat) 361634
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.