Novus Biologicals
Manufacturer Code:NBP200600
Catalog # NBP200600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H3 |
Immunogen | Full length human recombinant protein of human CCNB1IP1 (NP_878269) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: chromosome 14 open reading frame 18 cyclin B1 interacting protein 1 cyclin B1 interacting protein 1 E3 ubiquitin protein ligase E3 ubiquitin-protein ligase CCNB1IP1 EC 6.3.2.- enhancer of invasion 10 HEI10C14orf18Cyclin-B1-interacting protein 1 Human enhancer of invasion 10; cyclin B1 interacting protein 1, E3 ubiquitin protein ligase; Cyclin-B1-interacting protein 1; E3 ubiquitin-protein ligase CCNB1IP1; Human enhancer of invasion 10; RING-type E3 ubiquitin transferase CCNB1IP1
Gene Aliases: C14orf18; CCNB1IP1; HEI10
UniProt ID: (Human) Q9NPC3
Entrez Gene ID: (Human) 57820
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.