OriGene
Manufacturer Code:CF502700
Catalog # 50-166-1566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H3 |
Immunogen | Full length human recombit protein of human CCNB1IP1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C14orf18 HEI10; cyclin B1 interacting protein 1, E3 ubiquitin protein ligase; Cyclin-B1-interacting protein 1; E3 ubiquitin-protein ligase CCNB1IP1; Human enhancer of invasion 10; RING-type E3 ubiquitin transferase CCNB1IP1
Gene Aliases: C14orf18; CCNB1IP1; HEI10
UniProt ID: (Human) Q9NPC3
Entrez Gene ID: (Human) 57820
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.