Novus Biologicals
Manufacturer Code:NBP290018B
Catalog # NB068965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 63 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human CCL7/MCP-3/MARC (NP_006264.2 Gln24-Leu99). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 7; C-C motif chemokine 7 chemokine (C-C motif) ligand 7 FIC MARC MCP3Monocyte chemotactic protein 3 MCP-3Small-inducible cytokine A7 MGC138465 Monocyte chemoattractant protein 3 NC28SCYA6 SCYA7MGC138463 small inducible cytokine A7 (monocyte chemotactic protein 3); chemokine (C-C motif) ligand 7; MCP-3; Monocyte chemoattractant protein 3; Monocyte chemotactic protein 3; NC28; small inducible cytokine A7 (monocyte chemotactic protein 3); Small-inducible cytokine A7
Gene Aliases: CCL7; FIC; MARC; MCP-3; MCP3; NC28; SCYA6; SCYA7
UniProt ID: (Human) P80098
Entrez Gene ID: (Human) 6354
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.