R&D Systems
Manufacturer Code:MAB278SP
Catalog # MAB278SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 21445 |
Immunogen | <EM>E. coli</EM>-derived recombinant human CCL5/RANTES |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta-chemokine RANTES; C-C motif chemokine 5; chemokine (C-C motif) ligand 5; chemokine (C-C motif) ligand 5 D17S136Enormally T-expressed and presumably secreted EoCP Eosinophil chemotactic cytokine RANTES SISd SIS-delta small inducible cytokine A5 (RANTES) small inducible cytokine subfamily A (Cys-Cys) member 5 Small-inducible cytokine A5 T cell-specific protein P228 T-cell specific protein p288 TCP228T-cell-specific protein RANTES; EoCP; Eosinophil chemotactic cytokine; RANTES(3-68); RANTES(4-68); regulated upon activation, normally T-expressed, and presumably secreted; SIS-delta; small inducible cytokine subfamily A (Cys-Cys), member 5; Small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228
Gene Aliases: CCL5; D17S136E; eoCP; RANTES; SCYA5; SIS-delta; SISd; TCP228
UniProt ID: (Human) P13501
Entrez Gene ID: (Human) 6352
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.