Invitrogen
Manufacturer Code:MA523709
Catalog # PIMA523709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 21418 |
Immunogen | E. coli-derived recombit human CCL5/RANTES |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta-chemokine RANTES; C-C motif chemokine 5; C-C motif chemokine 5 SIS-delta T-cell specific protein p288 T-cell-specific protein RANTES beta-chemokine RANTES eoCP eosinophil chemotactic cytokine small inducible cytokine A5 t cell-specific protein P228 D17S136E MGC17164 SCYA5 SISd TCP228; chemokine (C-C motif) ligand 5; EoCP; Eosinophil chemotactic cytokine; RANTES(3-68); RANTES(4-68); regulated upon activation, normally T-expressed, and presumably secreted; SIS-delta; small inducible cytokine subfamily A (Cys-Cys), member 5; Small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228
Gene Aliases: CCL5; D17S136E; eoCP; RANTES; SCYA5; SIS-delta; SISd; TCP228
UniProt ID: (Human) P13501
Entrez Gene ID: (Human) 6352
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.