Invitrogen
Manufacturer Code:PA585254
Catalog # PIPA585254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human RANTES. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta-chemokine RANTES; C-C motif chemokine 5; chemokine (C-C motif) ligand 5; D17S136E antibody MGC17164 antibody RANTES antibody SCYA5 antibody SISd antibody TCP228 antibody CCL5 antibody small inducible cytokine subfamily A (Cys-Cys) member 5 antibody T-cell specific protein p288 antibody small inducible cytokine A5 (RANTES) antibody t cell-specific protein P228 antibody eosinophil chemotactic cytokine antibody eoCP antibody T-cell-specific protein RANTES antibody SIS-delta antibody regulated upon activation normally T-expressed and presumably secreted antibody beta-chemokine RANTES antibody C-C motif chemokine 5 antibody small-inducible cytokine A5 antibody chemokine (C-C motif) ligand 5 antibody; EoCP; Eosinophil chemotactic cytokine; RANTES(3-68); RANTES(4-68); regulated upon activation, normally T-expressed, and presumably secreted; SIS-delta; small inducible cytokine subfamily A (Cys-Cys), member 5; Small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228
Gene Aliases: CCL5; D17S136E; eoCP; RANTES; SCYA5; SIS-delta; SISd; TCP228
UniProt ID: (Human) P13501
Entrez Gene ID: (Human) 6352
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.