Invitrogen
Manufacturer Code:PA547536
Catalog # PIPA547536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit feline CCL5/RANTES Tyr26-Asn92 (Lys84Tyr and Gly85 del) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 5; C-C motif chemokine 5 SIS-delta T-cell specific protein p288 T-cell-specific protein RANTES beta-chemokine RANTES eoCP eosinophil chemotactic cytokine small inducible cytokine A5 t cell-specific protein P228 D17S136E MGC17164 SCYA5 SISd TCP228; small inducible cytokine A5; Small-inducible cytokine A5; T-cell-specific protein RANTES
Gene Aliases: CCL5; RANTES; SCYA5
UniProt ID: (Cat) Q8SQ40
Entrez Gene ID: (Cat) 493689
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.