R&D Systems
Manufacturer Code:MAB451500
Catalog # MAB451500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 46907 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse CCL4/MIP-1 beta Ala24-Asn92 Accession # Q5QNV9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ACT-2; ACT2 ACT-2 AT744.1 C-C motif chemokine 4 chemokine (C-C motif) ligand 4 Exodus-3 G-26 G-26 T-lymphocyte-secreted protein HC21 LAG1 LAG-1 Lymphocyte activation gene 1 protein Macrophage inflammatory protein 1-beta MIP1 beta MIP-1 beta MIP1B MIP1B1 MIP-1-beta MIP-1-beta(1-69) PAT 744 Protein H400 SCYA2 SCYA4 secreted protein G-26 SIS-gamma small inducible cytokine A4 (homologous to mouse Mip-1b) Small-inducible cytokine A4 T-cell activation protein 2; C-C motif chemokine 4; chemokine (C-C motif) ligand 4; G-26 T-lymphocyte-secreted protein; HC21; LAG-1; Lymphocyte activation gene 1 protein; Macrophage inflammatory protein 1-beta; MIP-1-beta; MIP-1-beta(1-69); MIP-1-beta(3-69); PAT 744; Protein H400; secreted protein G-26; SIS-gamma; small inducible cytokine A4 (homologous to mouse Mip-1b); Small-inducible cytokine A4; T-cell activation protein 2
Gene Aliases: ACT2; AT744.1; CCL4; G-26; HC21; LAG-1; LAG1; MIP-1-beta; MIP1B; MIP1B1; SCYA2; SCYA4
UniProt ID: (Human) P13236
Entrez Gene ID: (Human) 6351
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.