Novus Biologicals
Manufacturer Code:NBP177897
Catalog # NBP177897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant protein raised in yeast corresponding to the 69 amino acids of the mature porcine CCL4L1 beta protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACT-2; ACT2 ACT-2 AT744.1 C-C motif chemokine 4 chemokine (C-C motif) ligand 4 G-26 G-26 T-lymphocyte-secreted protein HC21 LAG1 LAG-1 Lymphocyte activation gene 1 protein Macrophage inflammatory protein 1-beta MIP-1 beta/CCL4 MIP1B MIP1B1 MIP-1-beta MIP-1-beta(1-69) PAT 744 Protein H400 SCYA2 SCYA4 secreted protein G-26 SIS-gamma small inducible cytokine A4 (homologous to mouse Mip-1b) Small-inducible cytokine A4 T-cell activation protein 2; C-C motif chemokine 4; chemokine (C-C motif) ligand 4; G-26 T-lymphocyte-secreted protein; HC21; LAG-1; Lymphocyte activation gene 1 protein; Macrophage inflammatory protein 1-beta; MIP-1-beta; MIP-1-beta(1-69); MIP-1-beta(3-69); PAT 744; Protein H400; secreted protein G-26; SIS-gamma; small inducible cytokine A4 (homologous to mouse Mip-1b); Small-inducible cytokine A4; T-cell activation protein 2
Gene Aliases: ACT2; AT744.1; CCL4; G-26; HC21; LAG-1; LAG1; MIP-1-beta; MIP1B; MIP1B1; SCYA2; SCYA4
UniProt ID: (Human) P13236
Entrez Gene ID: (Human) 6351
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.