Invitrogen
Manufacturer Code:PA547785
Catalog # PIPA547785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit canine CCL4/MIP-1 beta Ala24-Asn92 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 4; C-C motif ligand 4 macrophage inflammatory protein 1-beta MIP1-beta MIP1B MIP1B1 ACT2 AT744.1 G-26 HC21 lymphocyte-activation gene 1 LAG-1 LAG1 SCYA2 SCYA4; CC chemokine ligand 4; Small-inducible cytokine A4
Gene Aliases: CCL4; SCYA4
UniProt ID: (Dog) Q68AZ0
Entrez Gene ID: (Dog) 448786
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.