Invitrogen
Manufacturer Code:PA5114961
Catalog # PIPA5114961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CCL4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACT-2; ACT2; ACT2 ACT-2 AT744.1 CC chemokine ligand 4 C-C motif chemokine 4 C-C motif chemokine ligand 4 CCL4 chemokine (C-C motif) ligand 4 G-26 G-26 T-lymphocyte-secreted protein HC21 immune activation gene 2 immune activation protein 2 LAG1 LAG-1 Lymphocyte activation gene 1 protein Macrophage inflammatory protein 1-beta macrophage inflammatory protein-1 beta MGC104418 MGC126025 MGC126026 MIP-1 beta Mip1b MIP-1B Mip1-b MIP1B1 MIP1beta MIP-1BETA MIP-1-beta MIP1-beta MIP-1-beta(1-69) MIP-1-beta(3-69) PAT 744 Protein H400 putative MIP-1beta protein RP23-320E6.8 SCY4A SCYA2 SCYA4 secreted protein G-26 SIS-gamma small inducible cytokine A4 small inducible cytokine A4 (homologous to mouse Mip-1b) small-inducible cytokine A4 T-cell activation protein 2; C-C motif chemokine 4; chemokine (C-C motif) ligand 4; G-26 T-lymphocyte-secreted protein; HC21; Immune activation protein 2; LAG-1; Lymphocyte activation gene 1 protein; Macrophage inflammatory protein 1-beta; macrophage inflammatory protein-1 beta; MIP-1 beta; MIP-1-beta; MIP-1-beta(1-69); MIP-1-beta(3-69); PAT 744; Protein H400; secreted protein G-26; SIS-gamma; small inducible cytokine A4; small inducible cytokine A4 (homologous to mouse Mip-1b); Small-inducible cytokine A4; T-cell activation protein 2
Gene Aliases: Act-2; ACT2; AT744.1; CCL4; G-26; HC21; LAG-1; LAG1; MIP-1-beta; MIP-1B; Mip1-b; MIP1B; MIP1B1; SCYA2; SCYA4
UniProt ID: (Human) P13236, (Rat) P50230, (Mouse) P14097
Entrez Gene ID: (Human) 6351, (Rat) 116637, (Mouse) 20303
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.