Invitrogen
Manufacturer Code:PA129036
Catalog # PIPA129036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein - amino acids 1-69 of mature rat CCL4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACT-2; ACT2; C-C motif chemokine 4; Immune activation protein 2; Macrophage inflammatory protein 1-beta; macrophage inflammatory protein-1 beta; MIP-1 beta; MIP-1-beta; Protein H400; RP23-320E6.8 AT744.1 Act-2 MIP-1B Mip1b Scya4; SIS-gamma; small inducible cytokine A4; Small-inducible cytokine A4
Gene Aliases: Act-2; AT744.1; Ccl4; MIP-1B; Mip1-b; Mip1b; Scya4
UniProt ID: (Rat) P50230, (Mouse) P14097
Entrez Gene ID: (Rat) 116637, (Mouse) 20303
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.