Invitrogen
Manufacturer Code:11970642
Catalog # 50-112-2181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CR3M |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; LD78-alpha(4-69); Macrophage inflammatory protein 1-alpha; MIP-1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7 AI323804 G0S19-1 LD78alpha MIP-1alpha MIP1-(a) MIP1-alpha Mip1a Scya3; SIS-beta; small inducible cytokine A3 (homologous to mouse Mip-1a); Small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein
Gene Aliases: CCL3; G0S19-1; LD78ALPHA; MIP-1-alpha; MIP1A; SCYA3
UniProt ID: (Human) P10147
Entrez Gene ID: (Human) 6348
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.