R&D Systems
Manufacturer Code:MAB66252
Catalog # MAB66252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 769324 |
Immunogen | <EM>E. coli</EM>-derived recombinant rat CCL3/MIP-1 alpha Ala24-Ala92 Accession # P50229 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 3; C-C motif chemokine 3 MIP1-(a) AI323804 chemokine (C-C motif) ligand 3 G0S19-1 LD78a LD78alpha MIP1 alpha MIP-1 alpha Mip1a MIP-1alpha MIP1-alpha PAT 464.1 SCYA3 SIS-beta; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; LD78-alpha(4-69); Macrophage inflammatory protein 1-alpha; MIP-1-alpha; MIP-1-alpha(4-69); PAT 464.1; SIS-beta; small inducible cytokine A3 (homologous to mouse Mip-1a); Small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein
Gene Aliases: CCL3; G0S19-1; LD78ALPHA; MIP-1-alpha; MIP1A; SCYA3
UniProt ID: (Human) P10147
Entrez Gene ID: (Human) 6348
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.