R&D Systems
Manufacturer Code:IC450G
Catalog # IC450G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 39624 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse CCL3/MIP-1 alpha Ala24-Ala92 Accession # P10855 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 3; C-C motif chemokine 3 MIP1-(a) AI323804 chemokine (C-C motif) ligand 3 G0S19-1 LD78a LD78alpha MIP1 alpha MIP-1 alpha Mip1a MIP-1alpha MIP1-alpha PAT 464.1 SCYA3 SIS-beta; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; LD78-alpha(4-69); Macrophage inflammatory protein 1-alpha; MIP-1-alpha; MIP-1-alpha(4-69); PAT 464.1; SIS-beta; small inducible cytokine A3 (homologous to mouse Mip-1a); Small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein
Gene Aliases: CCL3; G0S19-1; LD78ALPHA; MIP-1-alpha; MIP1A; SCYA3
UniProt ID: (Human) P10147
Entrez Gene ID: (Human) 6348
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.