Novus Biologicals
Manufacturer Code:NBP28958550UL
Catalog # NB020654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 12 |
Immunogen | Recombinant Human CCL28 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 28; CC chemokine CCL28; chemokine (C-C motif) ligand 28 member 28 MGC71902 Protein CCK1; chemokine (C-C motif) ligand 28 splice variant chi; MEC; Mucosae-associated epithelial chemokine; Protein CCK1; small inducible cytokine A28; small inducible cytokine subfamily A (Cys-Cys), member 28; Small-inducible cytokine A28
Gene Aliases: CCK1; CCL28; MEC; SCYA28
UniProt ID: (Human) Q9NRJ3
Entrez Gene ID: (Human) 56477
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.