Invitrogen
Manufacturer Code:PA576665
Catalog # PIPA576665
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human CCL28. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 28; C-C motif chemokine 28CCK 1CCK1 proteinCCL 28CCL28CCL28&lowbarHUMANChemokine (C-C motif) ligand 28MECMucosae associated epithelial chemokineMucosae-associated epithelial chemokineProtein CCK1SCYA28Small inducible cytokine A28 &lsqbPrecursor]Small-inducible cytokine A28; CC chemokine CCL28; chemokine (C-C motif) ligand 28 splice variant chi; MEC; Mucosae-associated epithelial chemokine; Protein CCK1; small inducible cytokine A28; small inducible cytokine subfamily A (Cys-Cys), member 28; Small-inducible cytokine A28
Gene Aliases: CCK1; CCL28; MEC; SCYA28
UniProt ID: (Human) Q9NRJ3, (Mouse) Q9JIL2
Entrez Gene ID: (Human) 56477, (Mouse) 56838, (Rat) 114492
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.