Novus Biologicals
Manufacturer Code:NBP212111B
Catalog # NBP212111B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0177-7N28 |
Immunogen | Mouse TECK |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: C-C motif chemokine 25; C-C motif chemokine 25 chemokine (C-C motif) ligand 25 Chemokine TECK Ck beta-15 Ckb15 SCYA25TECKvar small inducible cytokine subfamily A (Cys-Cys) member 25 Small-inducible cytokine A25 TECKMGC150327 thymus expressed chemokine Thymus-expressed chemokine; chemokine (C-C motif) ligand 25; Chemokine TECK; Ck beta-15; small inducible cytokine subfamily A (Cys-Cys), member 25; Small-inducible cytokine A25; TECKvar; thymus expressed chemokine; Thymus-expressed chemokine
Gene Aliases: CCL25; Ckb15; SCYA25; TECK
UniProt ID: (Human) O15444
Entrez Gene ID: (Human) 6370
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.