Proteintech
Manufacturer Code:252851AP150UL
Catalog # 50-173-4836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CCL25/TECK Fusion Protein Ag17994 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C C motif chemokine 25 CCL25 Chemokine TECK Ckb15 SCYA25 Small inducible cytokine A25 TECK Thymus expressed chemokine; C-C motif chemokine 25; chemokine (C-C motif) ligand 25; Chemokine TECK; Ck beta-15; small inducible cytokine subfamily A (Cys-Cys), member 25; Small-inducible cytokine A25; TECKvar; thymus expressed chemokine; Thymus-expressed chemokine
Gene Aliases: CCL25; Ckb15; SCYA25; TECK
UniProt ID: (Human) O15444
Entrez Gene ID: (Human) 6370
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.