R&D Systems
Manufacturer Code:MAB343SP
Catalog # MAB343SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 61016 |
Immunogen | <EM>E. coli</EM>-derived recombinant human CCL24/Eotaxin-2/MPIF-2 Val27-Cys119 Accession # AAB51135 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 24; chemokine (C-C motif) ligand 24; chemokine (C-C motif) ligand 24 Ckb-6 Eosinophil chemotactic protein 2 Eotaxin-2 member 24 MPIF2 MPIF-2 MPIF2Myeloid progenitor inhibitory factor 2 MPIF-2Small-inducible cytokine A24 SCYA24CK-beta-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin-2; MPIF-2; Myeloid progenitor inhibitory factor 2; small inducible cytokine subfamily A (Cys-Cys), member 24; Small-inducible cytokine A24
Gene Aliases: CCL24; Ckb-6; MPIF-2; MPIF2; SCYA24
UniProt ID: (Human) O00175
Entrez Gene ID: (Human) 6369
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.