Proteintech
Manufacturer Code:223061AP150UL
Catalog # 50-173-3824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C C motif chemokine 24 CCL24 CK beta 6 Ckb 6 Eotaxin 2 MPIF 2 MPIF2 SCYA24 Small inducible cytokine A24; C-C motif chemokine 24; CC chemokine CCL24; chemokine (C-C motif) ligand 24; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin-2; MPIF-2; Myeloid progenitor inhibitory factor 2; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; Small-inducible cytokine A24
Gene Aliases: CCL24; CKb-6; MPIF-2; MPIF2; SCYA24
UniProt ID: (Human) O00175, (Mouse) Q9JKC0
Entrez Gene ID: (Human) 6369, (Mouse) 56221
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.