Invitrogen
Manufacturer Code:PA5100686
Catalog # PIPA5100686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CCL23 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 23; C6 beta-chemokine; C6 beta-chemokine C-C motif chemokine 23 C-C motif chemokine ligand 23 CCL23 CCL23(19-99) CCL23(22-99) CCL23(27-99) CCL23(30-99) chemokine (C-C motif) ligand 23 Ck beta 8-1 CKb8 Ckb-8 Ckb-8-1 CK-beta-8 hmrp-2a Macrophage inflammatory protein 3 MIP3 MIP-3 MIP-3CK-BETA-8 MPIF1 MPIF-1 MPIF-1Ckb-8-1 Myeloid progenitor inhibitory factor 1 SCYA23 small inducible cytokine subfamily A (Cys-Cys) member 23 Small-inducible cytokine A23; CCL23(19-99); CCL23(22-99); CCL23(27-99); CCL23(30-99); chemokine (C-C motif) ligand 23; CK-beta-8; CKB-8; Macrophage inflammatory protein 3; MIP-3; MPIF-1; Myeloid progenitor inhibitory factor 1; small inducible cytokine subfamily A (Cys-Cys), member 23; Small-inducible cytokine A23
Gene Aliases: CCL23; CK-BETA-8; Ckb-8; Ckb-8-1; CKb8; hmrp-2a; MIP-3; MIP3; MPIF-1; MPIF1; SCYA23
UniProt ID: (Human) P55773
Entrez Gene ID: (Human) 6368
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.