Invitrogen
Manufacturer Code:PA523643
Catalog # PIPA523643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 23-52 amino acids from the central region of human CCL21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6Ckine; 6Ckine CKb9 ECL SCYA21 SLC TCA4 C-C motif chemokine 21 Efficient Chemoattractant for Lymphocytes beta chemokine exodus-2 beta-chemokine exodus-2 exodus-2 secondary lymphoid tissue chemokine secondary lymphoid-tissue chemokine small inducible cytokine subfamily A (Cys-Cys) member 21 sma; beta chemokine exodus-2; Beta-chemokine exodus-2; C-C motif chemokine 21; C-C motif chemokine 21a; CC chemokine 6Ckine-ser; CCL21-Ser; chemokine (C-C motif) ligand 21; Efficient Chemoattractant for Lymphocytes; exodus-2; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine A21a; small inducible cytokine subfamily A (Cys-Cys), member 21; Small-inducible cytokine A21; Small-inducible cytokine A21a; TCA4; Thymus-derived chemotactic agent 4
Gene Aliases: 6CKBAC2; 6Ckine; ALP; AW987545; CCL21; Ccl21a; CKb9; ECL; plt; SCYA21; SCYA21a; Scya21b; SLC; TCA4; UNQ784/PRO1600
UniProt ID: (Human) O00585, (Mouse) P84444
Entrez Gene ID: (Human) 6366, (Mouse) 18829
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.