R&D Systems
Manufacturer Code:AF457SP
Catalog # AF457SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant mouse CCL21/6Ckine Ser24-Gly133 Accession # P84444 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 6Ckine; 6Ckine 6CkineSmall-inducible cytokine A21 Beta-chemokine exodus-2 chemokine (C-C motif) ligand 21 CKb9 exodus-2 member 21 SCYA21 SCYA21MGC34555 secondary lymphoid tissue chemokine SLC SLCSecondary lymphoid-tissue chemokine TCA-4; beta chemokine exodus-2; Beta-chemokine exodus-2; C-C motif chemokine 21; chemokine (C-C motif) ligand 21; Efficient Chemoattractant for Lymphocytes; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine subfamily A (Cys-Cys), member 21; Small-inducible cytokine A21
Gene Aliases: 6Ckine; CCL21; CKb9; ECL; SCYA21; SLC; TCA4; UNQ784/PRO1600
UniProt ID: (Human) O00585
Entrez Gene ID: (Human) 6366
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.