Novus Biologicals
Manufacturer Code:NBP227142
Catalog # NBP227142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 4N5F7 |
Immunogen | Full length recombinant mouse MIP-3 full length protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: beta chemokine exodus-1; beta chemokine exodus-1 Beta-chemokine exodus-1 CC chemokine LARC C-C motif chemokine 20 chemokine (C-C motif) ligand 20 CKb4 exodus-1 LARCLiver and activation-regulated chemokine MIP-3a MIP-3-alpha MIP3AMacrophage inflammatory protein 3 alpha SCYA20Small-inducible cytokine A20 small inducible cytokine subfamily A (Cys-Cys) member 20 ST38; Beta-chemokine exodus-1; C-C motif chemokine 20; CC chemokine LARC; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; Liver and activation-regulated chemokine; Macrophage inflammatory protein 3 alpha; MIP-3-alpha; small inducible cytokine subfamily A (Cys-Cys), member 20; Small-inducible cytokine A20
Gene Aliases: CCL20; CKb4; Exodus; LARC; MIP-3-alpha; MIP-3a; MIP3A; SCYA20; ST38
UniProt ID: (Human) P78556
Entrez Gene ID: (Human) 6364
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.