Abnova
Manufacturer Code:MAB1314
Catalog # 89-110-351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | mxsghk-3 |
Immunogen | Recombinant protein corresponding to full length human CCL20. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant CCL20.
For Research Use Only
Protein Aliases: beta chemokine exodus-1; Beta-chemokine exodus-1; C-C motif chemokine 20; CC chemokine LARC; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; LARC; Liver and activation-regulated chemokine; Macrophage inflammatory protein 3 alpha; MIP-3-alpha; MIP-3a; MIP3A; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; Small-inducible cytokine A20; ST38
Gene Aliases: CCL20; CKb4; Exodus; LARC; MIP-3-alpha; MIP-3a; MIP3A; SCYA20; ST38
UniProt ID: (Human) P78556
Entrez Gene ID: (Human) 6364
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.