Novus Biologicals
Manufacturer Code:NBP107034H
Catalog # NBP107034H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot# P14844] |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 2; C-C motif chemokine 2 chemokine (C-C motif) ligand 2 GDCF-2 HC11Monocyte secretory protein JE MCAFMonocyte chemotactic and activating factor MCP1 Monocyte chemotactic protein 1 MCP-1 Small-inducible cytokine A2 MGC9434 Monocyte chemoattractant protein 1 monocyte chemoattractant protein-1 SCYA2HSMCR30 small inducible cytokine A2 (monocyte chemotactic protein 1 homologous tomouse Sig-je) small inducible cytokine subfamily A (Cys-Cys) member 2 SMC-CF; chemokine (C-C motif) ligand 2; HC11; MCAF; MCP-1; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; Monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; Monocyte secretory protein JE; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; Small-inducible cytokine A2
Gene Aliases: CCL2; GDCF-2; HC11; HSMCR30; MCAF; MCP-1; MCP1; SCYA2; SMC-CF
UniProt ID: (Human) P13500
Entrez Gene ID: (Human) 6347
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.