Invitrogen
Manufacturer Code:14709181
Catalog # 50-136-22
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 4E2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 2; GDCF-2 HC11 HSMCR30 MCAF MCP-1 MCP1 MGC9434 SCYA2 SMC-CF; MCP-1; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; Monocyte chemotactic protein 1; Platelet-derived growth factor-inducible protein JE; small inducible cytokine A2; Small-inducible cytokine A2
Gene Aliases: AI323594; Ccl2; HC11; JE; MCAF; MCP-1; MCP1; Scya2; Sigje; SMC-CF
UniProt ID: (Mouse) P10148
Entrez Gene ID: (Mouse) 20296
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.