Invitrogen
Manufacturer Code:167096025
Catalog # 16-709-6025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 2H5 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: Acidic seminal fluid protein AI323594 C-C motif chemokine 2 C-C motif chemokine ligand 2 Ccl2 chemokine (C-C motif) ligand 2 GDCF-2 HC11 H-MCP-1 HSMCR30 immediate-early serum-responsive JE protein immediate-early serum-responsive protein JE Je MCAF MCP1 MCP-1 MCP1A MCP-1A MGC9434 Monocyte chemoattractant protein 1 monocyte chemoattractant protein-1 monocyte chemotactic and activating factor Monocyte chemotactic protein 1 monocyte chemotactic protein 1A Monocyte secretory protein JE monocyte-chemoattractant protein-1 precursor platelet-derived growth factor-inducible protein JE RP23-350G1.3 Scya2 Sigje small inducible cytokine A2 small inducible cytokine A2 (monocyte chemotactic protein 1 homologous to mouse Sig-je) small inducible cytokine subfamily A (Cys-Cys) member 2 small inducible gene JE small-inducible cytokine A2 SMC-CF; C-C motif chemokine 2; chemokine (C-C motif) ligand 2; HC11; immediate-early serum-responsive JE protein; Immediate-early serum-responsive protein JE; MCAF; MCP-1; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; Monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; Monocyte secretory protein JE; Platelet-derived growth factor-inducible protein JE; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; Small-inducible cytokine A2
Gene Aliases: AI323594; CCL2; GDCF-2; HC11; HSMCR30; JE; MCAF; MCP-1; MCP1; SCYA2; Sigje; SMC-CF
UniProt ID: (Human) P13500, (Rat) P14844, (Mouse) P10148
Entrez Gene ID: (Human) 6347, (Rat) 24770, (Mouse) 20296
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.