R&D Systems
Manufacturer Code:MAB7800SP
Catalog # MAB7800SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 866202 |
Immunogen | <EM>E. coli</EM>-derived recombinant rat CCL19/MIP-3 beta Gly26-Ser108 Accession # NP_001102131 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta chemokine exodus-3; beta chemokine exodus-3 Beta-chemokine exodus-3 CC chemokine ligand 19 C-C motif chemokine 19 chemokine (C-C motif) ligand 19 CKb11 EBI1-ligand chemokine ELC ELCMIP-3-beta Epstein-Barr virus-induced molecule 1 ligand chemokine exodus-3 Macrophage inflammatory protein 3 beta macrophage inflammatory protein 3-beta MGC34433 MIP3 beta MIP-3 beta MIP-3b MIP3BCK beta-11 SCYA19EBI1 ligand chemokine small inducible cytokine subfamily A (Cys-Cys) member 19 Small-inducible cytokine A19; Beta-chemokine exodus-3; C-C motif chemokine 19; CC chemokine ligand 19; chemokine (C-C motif) ligand 19; CK beta-11; EBI1 ligand chemokine; EBI1-ligand chemokine; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; Macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP-3-beta; small inducible cytokine subfamily A (Cys-Cys), member 19; Small-inducible cytokine A19
Gene Aliases: CCL19; CKb11; ELC; MIP-3b; MIP3B; SCYA19
UniProt ID: (Human) Q99731
Entrez Gene ID: (Human) 6363
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.