OriGene
Manufacturer Code:TA804426
Catalog # 50-168-0243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 22-98 of human CCL19 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta chemokine exodus-3; Beta-chemokine exodus-3; C-C motif chemokine 19; CC chemokine ligand 19; chemokine (C-C motif) ligand 19; CK beta-11; CKb11 ELC MIP-3b MIP3B SCYA19; EBI1 ligand chemokine; EBI1-ligand chemokine; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; Macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP-3-beta; small inducible cytokine subfamily A (Cys-Cys), member 19; Small-inducible cytokine A19
Gene Aliases: CCL19; CKb11; ELC; MIP-3b; MIP3B; SCYA19
UniProt ID: (Human) Q99731
Entrez Gene ID: (Human) 6363
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.