R&D Systems
Manufacturer Code:BAF420
Catalog # BAF420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse CCL11/Eotaxin (R&D Systems Catalog # 420-ME) His24-Pro97 Accession # P48298 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 11; C-C motif chemokine 11 chemokine (C-C motif) ligand 11 Eosinophil chemotactic protein Eotaxin eotaxin-1 MGC22554 SCYA11Small-inducible cytokine A11 small inducible cytokine subfamily A (Cys-Cys) member 11 (eotaxin); chemokine (C-C motif) ligand 11; Eosinophil chemotactic protein; Eotaxin; eotaxin-1; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); Small-inducible cytokine A11
Gene Aliases: CCL11; SCYA11
UniProt ID: (Human) P51671
Entrez Gene ID: (Human) 6356
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.