Invitrogen
Manufacturer Code:PA577377
Catalog # PIPA577377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CETPRIRGTGTRELE corresponding to amino acid residues 39-53 of mouse Cholecystokinin B Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK(B) receptor; CCK-B receptor; CCK-BR; CCK2 receptor; CCK2-R; CCK2R Gastrin Receptor; Cholecystokinin-2 receptor; gastrin receptor; Gastrin/cholecystokinin type B receptor
Gene Aliases: CCK-B; CCK-BR; CCK2-R; CCK2R; CCKBR; CCKR-2; CCKRB; Cholrec; GASR
UniProt ID: (Human) P32239, (Rat) P30553, (Mouse) P56481
Entrez Gene ID: (Human) 887, (Rat) 25706, (Mouse) 12426
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.