Invitrogen
Manufacturer Code:PA527169
Catalog # PIPA527169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 271 and 363 of Human CCK-BR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-B CCK2R GASR CCK-B receptor CCK-BR CCK2 receptor CCK2-R cholecystokinin-2 receptor gastrin receptor gastrin/cholecystokinin type B receptor; CCK-B receptor; CCK-BR; CCK2 receptor; CCK2-R; Cholecystokinin-2 receptor; gastrin receptor; Gastrin/cholecystokinin type B receptor
Gene Aliases: CCK-B; CCK2R; CCKBR; CCKRB; GASR
UniProt ID: (Human) P32239
Entrez Gene ID: (Human) 887
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.